siRNA Details
| Gene Symbol: LOC153684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015447.1 | 1 | 565 | |||
| GenBank mRNA | AK123995.1 | - | 1 | 606 | ||
| BC101382.1 | - | 1 | 566 | |||
| BC101383.2 | - | 1 | 566 | |||
| BC101384.1 | - | 1 | 565 | |||
| BC101385.2 | - | 1 | 565 | |||

LOC153684
Genomic Map