siRNA Details
| Gene Symbol: LINC00937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024420.1 | 2 | 633 | |||
| Gene Symbol: LOC105374312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017007611.1 | XP_016863100.1 | 4 | 844 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI909919.1 | - | 2 | 634 | Chr. 12 - 8356963 - 8390752 [-] on Build GRCh38 | |

LINC00937
Genomic Map