siRNA Details
| Gene Symbol: PQLC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024016.2 | 3 | 616 | |||
| GenBank mRNA | BE905784.1 | - | 3 | 615 | ||
| BX113701.1 | - | 3 | 587 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PQLC2L| Gene Symbol: PQLC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024016.2 | 3 | 616 | |||
| GenBank mRNA | BE905784.1 | - | 3 | 615 | ||
| BX113701.1 | - | 3 | 587 | |||