siRNA Details
Gene Symbol: GAS2L1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002942.1 | 3 | 1790 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GAS2L1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002942.1 | 3 | 1790 |