siRNA Details
| Gene Symbol: LINC00301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026946.1 | 7 | 743 | |||
| GenBank mRNA | AK058123.1 | - | 7 | 743 | ||
| BC029583.1 | - | 6 | 626 | |||
| BX103095.1 | - | 6 | 629 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI463377.1 | - | 6 | 631 | Chr. 11 - 60615769 - 60687149 [+] on Build GRCh38 | |

LINC00301
Genomic Map