siRNA Details
| Gene Symbol: PWRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026647.1 | 1 | 1634 | |||
| GenBank mRNA | AK124131.1 | - | 2 | 799 | ||
| BC032951.1 | - | 2 | 760 | |||
| BC070120.1 | - | 2 | 755 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PWRN2| Gene Symbol: PWRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026647.1 | 1 | 1634 | |||
| GenBank mRNA | AK124131.1 | - | 2 | 799 | ||
| BC032951.1 | - | 2 | 760 | |||
| BC070120.1 | - | 2 | 755 | |||