siRNA Details
Gene Symbol: GEMIN8P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002830.1 | 1 | 1641 | |||
GenBank mRNA | AY726571.1 | - | 1 | 1632 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GEMIN8P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002830.1 | 1 | 1641 | |||
GenBank mRNA | AY726571.1 | - | 1 | 1632 |