siRNA Details
| Gene Symbol: LOC152225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026934.1 | 1 | 13 | |||
| GenBank mRNA | AK090588.1 | - | 1 | 13 | ||
| BG249468.1 | - | 1 | 14 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC152225| Gene Symbol: LOC152225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026934.1 | 1 | 13 | |||
| GenBank mRNA | AK090588.1 | - | 1 | 13 | ||
| BG249468.1 | - | 1 | 14 | |||