siRNA Details
| Gene Symbol: MRGPRG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027138.1 | 2 | 251 | |||
| GenBank mRNA | AK093421.1 | - | 2 | 251 | ||
| AK097749.1 | - | 3 | 284 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRGPRG-AS1| Gene Symbol: MRGPRG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027138.1 | 2 | 251 | |||
| GenBank mRNA | AK093421.1 | - | 2 | 251 | ||
| AK097749.1 | - | 3 | 284 | |||