siRNA Details
Gene Symbol: MRGPRG-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027138.1 | 2 | 1494 | |||
GenBank mRNA | AK093421.1 | - | 2 | 1493 | ||
AK097749.1 | - | 3 | 1526 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MRGPRG-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027138.1 | 2 | 1494 | |||
GenBank mRNA | AK093421.1 | - | 2 | 1493 | ||
AK097749.1 | - | 3 | 1526 |