siRNA Details
| Gene Symbol: MRGPRG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027138.1 | 2 | 829 | |||
| GenBank mRNA | AK093421.1 | - | 2 | 829 | ||
| AK097749.1 | - | 3 | 862 | |||
| AY422795.1 | - | 1 | 496 | |||
| BC104971.1 | - | 1 | 534 | |||
| BC104973.1 | - | 1 | 534 | |||

MRGPRG-AS1
Genomic Map