siRNA Details
| Gene Symbol: ZNF702P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003578.1 | 4 | 1976 | |||
| GenBank mRNA | AK023047.1 | - | 1 | 1189 | ||
| AK092593.1 | - | 4 | 1976 | |||
| AL832505.1 | - | 4 | 1973 | |||
| BC032590.1 | - | 1 | 1355 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF702P| Gene Symbol: ZNF702P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003578.1 | 4 | 1976 | |||
| GenBank mRNA | AK023047.1 | - | 1 | 1189 | ||
| AK092593.1 | - | 4 | 1976 | |||
| AL832505.1 | - | 4 | 1973 | |||
| BC032590.1 | - | 1 | 1355 | |||