siRNA Details
| Gene Symbol: LOC283332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026948.1 | 2 | 131 | |||
| GenBank mRNA | AK093215.1 | - | 2 | 125 | ||
| BC121095.1 | - | 1 | 78 | |||
| BC121096.1 | - | 1 | 78 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB039238.1 | - | 2 | 127 | Chr. 12 - 49910760 - 49911858 [-] on Build GRCh38 | |

LOC283332
Genomic Map