siRNA Details
Gene Symbol: LOC283332 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026948.1 | 3 | 2305 | |||
GenBank mRNA | AK093215.1 | - | 3 | 2299 | ||
BX092566.1 | - | 2 | 246 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC283332 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026948.1 | 3 | 2305 | |||
GenBank mRNA | AK093215.1 | - | 3 | 2299 | ||
BX092566.1 | - | 2 | 246 |