siRNA Details
| Gene Symbol: LOC389834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027420.1 | 1 | 1113 | |||
| GenBank mRNA | AK123403.1 | - | 1 | 1113 | ||
| BC137367.1 | - | 1 | 957 | 9021365 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC389834| Gene Symbol: LOC389834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027420.1 | 1 | 1113 | |||
| GenBank mRNA | AK123403.1 | - | 1 | 1113 | ||
| BC137367.1 | - | 1 | 957 | 9021365 | ||