siRNA Details
| Gene Symbol: LINC01128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047519.1 | 6 | 1483 | |||
| NR_047521.1 | 5 | 1330 | ||||
| NR_047523.1 | 4 | 1234 | ||||
| NR_047524.1 | 4 | 1146 | ||||
| NR_047525.1 | 5 | 1197 | ||||
| GenBank mRNA | AK026873.1 | - | 7 | 1081 | ||
| AK311732.1 | - | 6 | 1184 | |||
| BC042880.1 | - | 5 | 1312 | |||

LINC01128
Genomic Map