siRNA Details
| Gene Symbol: FAM86EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130740.1 | 4 | 1335 | |||
| NR_130741.1 | 4 | 389 | ||||
| NR_130742.1 | 4 | 339 | ||||
| GenBank mRNA | BM718734.1 | - | 2 | 166 | ||
| CN481473.1 | - | 3 | 250 | |||
| HY065247.1 | - | 4 | 339 | |||

FAM86EP
Genomic Map