siRNA Details
| Gene Symbol: FAM86EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130740.1 | 4 | 1753 | |||
| NR_130741.1 | 4 | 807 | ||||
| NR_130742.1 | 4 | 757 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CD674875.1 | - | 3 | 612 | Chr. 4 - 3942852 - 3955363 [-] on Build GRCh38 | |

FAM86EP
Genomic Map