siRNA Details
Gene Symbol: TRHDE-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026836.1 | 1 | 98 | |||
NR_026837.1 | 1 | 98 | ||||
GenBank mRNA | BC040230.1 | - | 1 | 73 | ||
DA024500.1 | - | 1 | 98 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TRHDE-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026836.1 | 1 | 98 | |||
NR_026837.1 | 1 | 98 | ||||
GenBank mRNA | BC040230.1 | - | 1 | 73 | ||
DA024500.1 | - | 1 | 98 |