siRNA Details
| Gene Symbol: TRHDE-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026836.1 | 1 | 571 | |||
| NR_026837.1 | 1 | 571 | ||||
| GenBank mRNA | BC040230.1 | - | 1 | 546 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRHDE-AS1| Gene Symbol: TRHDE-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026836.1 | 1 | 571 | |||
| NR_026837.1 | 1 | 571 | ||||
| GenBank mRNA | BC040230.1 | - | 1 | 546 | ||