siRNA Details
| Gene Symbol: RNF138P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001575.1 | 1 | 2400 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF138P1| Gene Symbol: RNF138P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001575.1 | 1 | 2400 | |||