siRNA Details
| Gene Symbol: C21orf62-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024622.1 | 4 | 897 | |||
| NR_024623.1 | 4 | 885 | ||||
| GenBank mRNA | AK096601.1 | - | 4 | 886 | ||
| BC034794.1 | - | 2 | 611 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C21orf62-AS1| Gene Symbol: C21orf62-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024622.1 | 4 | 897 | |||
| NR_024623.1 | 4 | 885 | ||||
| GenBank mRNA | AK096601.1 | - | 4 | 886 | ||
| BC034794.1 | - | 2 | 611 | |||