siRNA Details
| Gene Symbol: CHODL-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024354.1 | 2 | 294 | |||
| GenBank mRNA | T74237.1 | - | 2 | 294 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | F12577.1 | - | 295 | Chr. - - [-] on Build | ||

CHODL-AS1
Genomic Map