siRNA Details
| Gene Symbol: LINC00111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024367.1 | 2, 3 | 360 | |||
| GenBank mRNA | BX099637.1 | - | 2, 3 | 496 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00111| Gene Symbol: LINC00111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024367.1 | 2, 3 | 360 | |||
| GenBank mRNA | BX099637.1 | - | 2, 3 | 496 | ||