siRNA Details
| Gene Symbol: LINC00628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027022.1 | 1 | 110 | |||
| GenBank mRNA | BC015457.1 | - | 1 | 110 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00628| Gene Symbol: LINC00628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027022.1 | 1 | 110 | |||
| GenBank mRNA | BC015457.1 | - | 1 | 110 | ||