siRNA Details
Gene Symbol: LINC00282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027047.1 | 5 | 2823 | |||
NR_027048.1 | 3 | 2680 | ||||
GenBank mRNA | AK094626.1 | - | 1 | 1369 | ||
AK124707.1 | - | 3 | 2680 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027047.1 | 5 | 2823 | |||
NR_027048.1 | 3 | 2680 | ||||
GenBank mRNA | AK094626.1 | - | 1 | 1369 | ||
AK124707.1 | - | 3 | 2680 |