siRNA Details
| Gene Symbol: AADACP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026915.1 | 3 | 270 | |||
| GenBank mRNA | BC014344.1 | - | 3 | 270 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE972233.1 | - | 3 | 271 | Chr. 3 - 151770456 - 151784237 [+] on Build GRCh38 | |

AADACP1
Genomic Map