siRNA Details
| Gene Symbol: AADACP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026915.1 | 3 | 1263 | |||
| GenBank mRNA | BC014344.1 | - | 3 | 1266 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AADACP1| Gene Symbol: AADACP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026915.1 | 3 | 1263 | |||
| GenBank mRNA | BC014344.1 | - | 3 | 1266 | ||