siRNA Details
Gene Symbol: FAM181A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027004.1 | 11 | 2226 | |||
GenBank mRNA | AK125743.1 | - | 3 | 864 | ||
BC037859.2 | - | 11 | 2226 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM181A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027004.1 | 11 | 2226 | |||
GenBank mRNA | AK125743.1 | - | 3 | 864 | ||
BC037859.2 | - | 11 | 2226 |