siRNA Details
| Gene Symbol: SNHG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001459.2 | 1 | 970 | |||
| NR_003138.3 | 1 | 970 | ||||
| GenBank mRNA | AK097876.1 | - | 1 | 831 | ||
| BC036576.2 | - | 1 | 725 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM701838.1 | - | 1 | 557 | Chr. 14 - 95533817 - 95534460 [-] on Build GRCh38 | |

SNHG10
Genomic Map