siRNA Details
Gene Symbol: SNHG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001459.2 | 1 | 874 | |||
NR_003138.3 | 1 | 874 | ||||
GenBank mRNA | AK097876.1 | - | 1 | 735 | ||
BC036576.2 | - | 1 | 629 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SNHG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001459.2 | 1 | 874 | |||
NR_003138.3 | 1 | 874 | ||||
GenBank mRNA | AK097876.1 | - | 1 | 735 | ||
BC036576.2 | - | 1 | 629 |