siRNA Details
| Gene Symbol: LINC00523 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024096.1 | 1 | 149 | |||
| GenBank mRNA | AK094097.1 | - | 1 | 167 | ||
| BX161445.1 | - | 1 | 149 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00523| Gene Symbol: LINC00523 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024096.1 | 1 | 149 | |||
| GenBank mRNA | AK094097.1 | - | 1 | 167 | ||
| BX161445.1 | - | 1 | 149 | |||