siRNA Details
| Gene Symbol: LINC00323 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024100.1 | 2 | 1586 | |||
| GenBank mRNA | AK094492.1 | - | 2 | 1586 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00323| Gene Symbol: LINC00323 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024100.1 | 2 | 1586 | |||
| GenBank mRNA | AK094492.1 | - | 2 | 1586 | ||