siRNA Details
| Gene Symbol: LINC00477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029451.2 | 2, 3 | 1342 | |||
| GenBank mRNA | AK057456.1 | - | 2, 3 | 1342 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ENSG00000197503
LINC00477| Gene Symbol: LINC00477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029451.2 | 2, 3 | 1342 | |||
| GenBank mRNA | AK057456.1 | - | 2, 3 | 1342 | ||