siRNA Details
| Gene Symbol: LOC652276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015441.1 | 2 | 660 | |||
| GenBank mRNA | AK096394.1 | - | 2 | 694 | ||
| AK291606.1 | - | 2 | 644 | |||
| BC062576.1 | - | 2 | 624 | |||
| BC080650.1 | - | 2 | 590 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC652276| Gene Symbol: LOC652276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015441.1 | 2 | 660 | |||
| GenBank mRNA | AK096394.1 | - | 2 | 694 | ||
| AK291606.1 | - | 2 | 644 | |||
| BC062576.1 | - | 2 | 624 | |||
| BC080650.1 | - | 2 | 590 | |||