siRNA Details
| Gene Symbol: LOC652276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015441.1 | 4 | 1593 | |||
| GenBank mRNA | AK096394.1 | - | 4 | 1627 | ||
| BQ214302.1 | - | 1 | 154 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC652276| Gene Symbol: LOC652276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015441.1 | 4 | 1593 | |||
| GenBank mRNA | AK096394.1 | - | 4 | 1627 | ||
| BQ214302.1 | - | 1 | 154 | |||