siRNA Details
| Gene Symbol: LINC00926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024433.1 | 3 | 2496 | |||
| GenBank mRNA | AK097083.1 | - | 3 | 2518 | ||
| AK123700.1 | - | 3 | 1894 | |||
| AL713736.1 | - | 1 | 1450 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00926| Gene Symbol: LINC00926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024433.1 | 3 | 2496 | |||
| GenBank mRNA | AK097083.1 | - | 3 | 2518 | ||
| AK123700.1 | - | 3 | 1894 | |||
| AL713736.1 | - | 1 | 1450 | |||