siRNA Details
| Gene Symbol: LINC00926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024433.1 | 2 | 1212 | |||
| GenBank mRNA | AK090515.1 | - | 1 | 1246 | ||
| AK097083.1 | - | 2 | 1212 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00926| Gene Symbol: LINC00926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024433.1 | 2 | 1212 | |||
| GenBank mRNA | AK090515.1 | - | 1 | 1246 | ||
| AK097083.1 | - | 2 | 1212 | |||