siRNA Details
| Gene Symbol: EWSAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026949.1 | 3 | 977 | |||
| GenBank mRNA | AK091087.1 | - | 3 | 965 | ||
| AK097560.1 | - | 4 | 806 | |||
| AK292174.1 | - | 3 | 530 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA999185.1 | - | 3 | 530 | Chr. 15 - 69080886 - 69091958 [+] on Build GRCh38 | |

EWSAT1
Genomic Map