siRNA Details
Gene Symbol: LOC341056 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027288.1 | 1 | 820 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC341056 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027288.1 | 1 | 820 |