siRNA Details
| Gene Symbol: GUCY2EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024042.1 | 21 | 2662 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GUCY2EP| Gene Symbol: GUCY2EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024042.1 | 21 | 2662 | |||