siRNA Details
| Gene Symbol: LOC283731 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027073.1 | 2 | 896 | |||
| GenBank mRNA | AK095530.1 | - | 2 | 793 | ||
| BC050067.1 | - | 1 | 796 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC283731| Gene Symbol: LOC283731 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027073.1 | 2 | 896 | |||
| GenBank mRNA | AK095530.1 | - | 2 | 793 | ||
| BC050067.1 | - | 1 | 796 | |||