siRNA Details
| Gene Symbol: LINC00189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027072.2 | 2 | 962 | |||
| GenBank mRNA | AF490769.1 | - | 4 | 637 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00189| Gene Symbol: LINC00189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027072.2 | 2 | 962 | |||
| GenBank mRNA | AF490769.1 | - | 4 | 637 | ||