siRNA Details
Gene Symbol: LINC00189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027072.2 | 2 | 886 | |||
GenBank mRNA | AF490769.1 | - | 4 | 561 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000420364.1 | - | 2 | 886 | Chr. 21 - 29193494 - 29288205 [+] on Build GRCh38 |
