siRNA Details
| Gene Symbol: CEP83-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027035.1 | 1 | 972 | |||
| GenBank mRNA | AK057269.1 | - | 1 | 972 | ||
| BC035086.2 | - | 1 | 955 | |||
| BC044616.1 | - | 1 | 956 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEP83-AS1| Gene Symbol: CEP83-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027035.1 | 1 | 972 | |||
| GenBank mRNA | AK057269.1 | - | 1 | 972 | ||
| BC035086.2 | - | 1 | 955 | |||
| BC044616.1 | - | 1 | 956 | |||