siRNA Details
| Gene Symbol: SLCO4A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024470.1 | 1 | 131 | |||
| GenBank mRNA | BC025345.1 | - | 1 | 131 | 4862156 | |
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG763914.1 | - | 1 | 132 | Chr. 20 - 62665215 - 62666622 [-] on Build GRCh38 | |

SLCO4A1-AS1
Genomic Map