siRNA Details
| Gene Symbol: GOLGA8IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024074.1 | 5 | 477 | |||
| GenBank mRNA | AK093104.1 | - | 5 | 477 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AI819685.1 | - | 1 | 39 | Chr. 15 - 22608052 - 22613809 [-] on Build GRCh38 | |

GOLGA8IP
Genomic Map