siRNA Details
| Gene Symbol: DSCR10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027695.1 | 3 | 465 | |||
| GenBank mRNA | AB066291.1 | - | 3 | 466 | ||
| BC093855.1 | - | 1 | 115 | 7939700 | ||
| BX115939.1 | - | 3 | 433 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DSCR10| Gene Symbol: DSCR10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027695.1 | 3 | 465 | |||
| GenBank mRNA | AB066291.1 | - | 3 | 466 | ||
| BC093855.1 | - | 1 | 115 | 7939700 | ||
| BX115939.1 | - | 3 | 433 | |||