siRNA Details
| Gene Symbol: HCG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002139.2 | 1 | 1462 | |||
| GenBank mRNA | AF036977.1 | - | 1 | 1460 | ||
| BC011879.1 | - | 1 | 1246 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HCG4| Gene Symbol: HCG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002139.2 | 1 | 1462 | |||
| GenBank mRNA | AF036977.1 | - | 1 | 1460 | ||
| BC011879.1 | - | 1 | 1246 | |||