siRNA Details
| Gene Symbol: LINC00593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026764.1 | 4 | 244 | |||
| GenBank mRNA | BC031958.1 | - | 4 | 244 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00593| Gene Symbol: LINC00593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026764.1 | 4 | 244 | |||
| GenBank mRNA | BC031958.1 | - | 4 | 244 | ||